Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phenoxathiin 98.0+%, TCI America™
SDP

Catalog No. P129625G Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
5 g
25 g

Chemical Identifiers

CAS 262-20-4
Molecular Formula C12H8OS
Molecular Weight (g/mol) 200.26
MDL Number MFCD00046933
InChI Key GJSGGHOYGKMUPT-UHFFFAOYSA-N
Synonym phenoxathiin, dibenzothioxin, dibenzooxathiane, phenothioxin, phenoxthin, phenoxathiane, phenoxathine, phenoxathrin, 1,4-dibenzothioxine, usaf do-17
PubChem CID 9217
IUPAC Name phenoxathiine
SMILES O1C2=CC=CC=C2SC2=CC=CC=C12

Specifications

Melting Point 57°C
Color White
Boiling Point 311°C
Quantity 25 g
Formula Weight 200.26
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Phenoxathiin
RTECSNumber SP6825000
TSCA Yes
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.