Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phenoxy resin, medium molecular weight, average M.W. 30000, pellets, Thermo Scientific™

Catalog No. AC177992500
missing translation for 'orderingAttributeHoverText'
Quantity:
250 g

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 26402-79-9

Specifications

Physical Form Pellet
Density 1.18g/mL
Quantity 250 g
Linear Formula {-C6H4-4-C(CH3)2C6H4-4-OCH2CH(OH)CH2O-}n
Chemical Name or Material Phenoxy resin, medium molecular weight, average M.W. 30000, pellets, medium molecular weight, average M.W. 30000
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.