Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phenoxyacetic acid, 98%

Catalog No. p-7020548
Click to view available options
Quantity:
100 g
500 g
2500 g

CAS: 122-59-8 | C8H8O3 | 152.15 g/mol

Phenoxyacetic acid is a flavoring ingredient. Its photodegradation using titanium dioxide as photocatalyst has been studied. Selective separation of penicillin V from phenoxyacetic acid using liquid membranes consisting of 1,2-dichloroethane and Amberlite LA-2 as carrier has been studied.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Phenoxyacetic acid is a flavoring ingredient. Its photodegradation using titanium dioxide as photocatalyst has been studied. Selective separation of penicillin V from phenoxyacetic acid using liquid membranes consisting of 1,2-dichloroethane and Amberlite LA-2 as carrier has been studied.

Solubility
Soluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 122-59-8
Molecular Formula C8H8O3
Molecular Weight (g/mol) 152.15
MDL Number MFCD00004296
InChI Key LCPDWSOZIOUXRV-UHFFFAOYSA-N
Synonym phenoxyacetic acid, phenoxyethanoic acid, acetic acid, phenoxy, glycolic acid phenyl ether, phenoxy acetic acid, glycol acid phenyl ether, o-phenylglycolic acid, phenoxyacetate, glycollic acid phenyl ether, acide phenoxyacetique
PubChem CID 19188
ChEBI CHEBI:8075
IUPAC Name 2-phenoxyacetic acid
SMILES OC(=O)COC1=CC=CC=C1

Specifications

Melting Point 98°C to 101°C
Boiling Point 285°C
Flash Point 165°C (329°F)
Quantity 100 g
Beilstein 907949
Merck Index 14,7254
Solubility Information Soluble in water.
Formula Weight 152.15
Percent Purity 99%
Chemical Name or Material Phenoxyacetic acid
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 204-556-7
RTECSNumber AJ2230000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.