Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phenoxyacetone 97.0+%, TCI America™
SDP

Catalog No. P011225G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P011225G 25 g
P01125G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P011225G Supplier TCI America Supplier No. P011225G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 621-87-4
Molecular Formula C9H10O2
Molecular Weight (g/mol) 150.18
MDL Number MFCD00008767
InChI Key QWAVNXZAQASOML-UHFFFAOYSA-N
Synonym phenoxyacetone, 2-propanone, 1-phenoxy, 1-phenoxy-2-propanone, 1-phenoxyacetone, 1-phenoxy-propan-2-one, phenoxy-2-propanone, phenoxymethyl methyl ketone, phenoxyaceton, phenoxy acetone, pubchem20265
PubChem CID 69313
IUPAC Name 1-phenoxypropan-2-one
SMILES CC(=O)COC1=CC=CC=C1

Specifications

Color Yellow
Boiling Point 230°C
Quantity 25 g
Formula Weight 150.18
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material Phenoxyacetone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.