Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Phenoxyethanol, Reagent, 99%, Spectrum™ Chemical

Click to view available options
Quantity:
120 mL
500 mL
1 L
4 L
20 L
4 x 4 L
200 L
Packaging:
Amber Glass Bottle
Description
Spectrum™ Chemical Phenoxyethanol, Reagent , is a glycol ether and an organic chemical compound. This colorless oily liquid is often used in dermalogical products and often replaces sodium azide as a biological buffer. It can also be found as a preservative in cosmetics, vaccines and various pharmaceuticals. Its reagent grade means this is the highest quality commercially available for this chemical and that the American Chemical Society has not officially set any specifications for this material.
Specifications
Specifications
CAS Max % | 1 |
Boiling Point | 244 to 250°C |
Packaging | Amber Glass Bottle |
Quantity | 20 L |
Percent Purity | 99% |
Grade | Reagent |
Physical Form | Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction