Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ Phentolamine Mesylate
GREENER_CHOICESDP

Catalog No. 6431100 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
64-311-00 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 64-311-00 Supplier Tocris Bioscience™ Supplier No. 6431/100

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Adrenergic α receptor antagonist; antihypertensive

Phentolamine Mesylate is a non-selective adrenergic α receptor antagonist (pKB = 8.07). Intravenous injection causes hypotension and tachycardia in rats. Intraperitoneal injection in mice increases plasma FGF21 levels.

Specifications

Chemical Name or Material 3-[[(4,5-Dihydro-1H-imidazol-2-yl)methyl](4-methylphenyl)amino]phenol methanesulfonate
CAS 65-28-1
Assay Percent Range >98%
Quantity 100 mg
Molecular Formula C17H19N3O.CH4O3S
Formula Weight Observed MW: 377.46
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.