Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phenyl alpha-D-Glucopyranoside 97.0+%, TCI America™
SDP

Catalog No. P13461G Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
1 g

Chemical Identifiers

CAS 4630-62-0
Molecular Formula C12H16O6
Molecular Weight (g/mol) 256.254
MDL Number MFCD00006594
InChI Key NEZJDVYDSZTRFS-ZIQFBCGOSA-N
Synonym Phenyl alpha-D-Glucoside
PubChem CID 99489
IUPAC Name (2R,3S,4S,5R,6R)-2-(hydroxymethyl)-6-phenoxyoxane-3,4,5-triol
SMILES C1=CC=C(C=C1)OC2C(C(C(C(O2)CO)O)O)O

Specifications

Melting Point 172°C
Color White-Yellow
Quantity 1 g
Formula Weight 256.25
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Phenyl alpha-D-Glucopyranoside
RTECSNumber LZ5985500
TSCA No
Recommended Storage Freezer
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.