Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Phenyl chlorothionocarbonate, 99%, ACROS Organics™
Click to view available options
Quantity:
1g
5g
25g
Packaging:
Glass bottle
Specifications
Color | Yellow |
Specific Gravity | 1.248 |
Boiling Point | 81.0°C to 83.0°C (6.0mmHg) |
Flash Point | 81°C |
Density | 1.2480g/mL |
Infrared Spectrum | Authentic |
Packaging | Glass bottle |
Refractive Index | 1.4795 to 1.4815 |
Quantity | 5g |
Assay Percent Range | 98.5% min. (GC) |
Show More |
Safety and Handling
GHS Signal Word: Danger
EINECSNumber : 213-736-4