Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phenyl disulfide, 99%

Catalog No. p-181906
Click to view available options
Quantity:
50 g
250 g
Packaging:
Plastic bottle

CAS: 882-33-7 | C12H10S2 | 218.33 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 882-33-7
Molecular Formula C12H10S2
Molecular Weight (g/mol) 218.33
MDL Number MFCD00003065
InChI Key GUUVPOWQJOLRAS-UHFFFAOYSA-N
Synonym diphenyl disulfide, phenyl disulfide, disulfide, diphenyl, diphenyldisulfide, 1,2-diphenyldisulfane, phenyldisulfanyl benzene, diphenyl disulphide, biphenyl disulfide, disulfide diphenyl, phenyldithiobenzene
PubChem CID 13436
IUPAC Name (phenyldisulfanyl)benzene
SMILES C1=CC=C(C=C1)SSC2=CC=CC=C2

Specifications

Melting Point 58°C to 61°C
Color White
Boiling Point 191°C to 192°C (15.0 mmHg)
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (HPLC)
Packaging Plastic bottle
Linear Formula (C6H5S-)2
Quantity 50 g
Beilstein 06, 323
Fieser 05,276; 06,235; 08,210; 10,171; 11,220; 12,514; 13,156
Merck Index 15, 3355
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in ether and alcohol
Formula Weight 218.33
Percent Purity 99%
Physical Form Crystalline Solid
Chemical Name or Material Phenyl disulfide, 99%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
Very toxic to aquatic life with long lasting effects.
Precautionary Statement GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and kee
EINECSNumber 212-926-4
RTECSNumber SS6825000
TSCA TSCA

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.