Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phenyl isothiocyanate, 98%

Catalog No. p-4510109
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

CAS: 103-72-0 | C7H5NS | 135.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 103-72-0
Molecular Formula C7H5NS
Molecular Weight (g/mol) 135.18
MDL Number MFCD00004798
InChI Key QKFJKGMPGYROCL-UHFFFAOYSA-N
Synonym phenyl isothiocyanate, phenylisothiocyanate, thiocarbanil, phenyl mustard oil, benzene, isothiocyanato, pitc, isothiocyanic acid phenyl ester, benzene-1-isothiocyanate, phenylsenfoel, phenyl thioisocyanate
PubChem CID 7673
ChEBI CHEBI:85103
IUPAC Name isothiocyanatobenzene
SMILES S=C=NC1=CC=CC=C1

Specifications

Melting Point -21.0°C
Color Yellow
Density 1.1288g/mL
Boiling Point 221.0°C
Flash Point 87°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.6490 to 1.6510
Linear Formula C6H5NCS
Quantity 100 g
Beilstein 12,453
Fieser 01,844; 02,323
Merck Index 15,7408
Specific Gravity 1.1288
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in ethanol and diethyl ether
Viscosity 1.3 mPa.s (20°C)
Formula Weight 135.19
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material Phenyl isothiocyanate
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes severe skin burns and eye damage.
May cause an allergic skin reaction.
Toxic in contact with skin.
Toxic if inhaled.
Toxic if swallowed.
Suspected of damaging the unborn child.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Immediately
EINECSNumber 203-138-1

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.