Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phenyl phosphorodiamidate, 97%, Thermo Scientific™

Catalog No. AC300440050
Click to view available options
:
5g; Glass bottle

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 7450-69-3
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Molecular Formula C6H9N2O2P
Quantity 5g
MDL Number MFCD00014767
Beilstein 06, IV, 751
Synonym phenyl phosphorodiamidate, phenylphosphorodiamidate, phenyl diamidophosphate, phosphorodiamidic acid, phenyl ester, diaminophosphoryl oxy benzene, phenylphosphorodiamidat, phosphoric phenyl ester diamide, phenyl phosphordiamidate, phenyl diamidophosphate #, diaminophenoxyphosphino-1-one
Solubility Information Solubility in water: soluble. Other solubilities: soluble in methanol,ethanol,chcl3,ccl4
InChI Key AYRRNFHDJUXLEQ-UHFFFAOYSA-N
SMILES C1=CC=C(C=C1)OP(=O)(N)N
IUPAC Name diaminophosphoryloxybenzene
Molecular Weight (g/mol) 172.12
PubChem CID 81954
Formula Weight 172.12
Percent Purity 97%
Chemical Name or Material Phenyl phosphorodiamidate, 97%
Show More Show Less
Hazard Category MOISTURE SENSITIVE
EINECSNumber 231-218-6
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.