Alfa Aesar™ Phenyl propargyl ether, 97%

Manufacturer: Alfa Aesar™ 4392303

Quantity 1g

 View more versions of this product

Catalog No. AA4392303

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 13610-02-1
    Molecular Formula C9H8O
    Linear Formula C6H5OCH2C≡CH
    Formula Weight 132.16
    MDL Number MFCD00048107
    Synonym 3-Phenoxy-1-propyne
    Chemical Name or Material Phenyl propargyl ether

    Specifications

    Formula Weight 132.16
    Health Hazard 3 GHS P Statement
    P210-P261-P280-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Molecular Formula C9H8O
    Linear Formula C6H5OCH2C≡CH
    Odor Pungent
    Physical Form Liquid
    MDL Number MFCD00048107
    CAS 13610-02-1
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H318-H335-H227-H315
    Causes serious eye damage.
    May cause respiratory irritation.
    Combustible liquid.
    Causes skin irritation.
    Refractive Index 1.5330
    Boiling Point 89° to 90°C (14mmHg)
    Color Colorless to yellow
    Synonym 3-Phenoxy-1-propyne
    Density 1.030g/mL
    Quantity 1g
    Chemical Name or Material Phenyl propargyl ether
    Assay 97%

    GHS H Statement
    H318-H335-H227-H315
    Causes serious eye damage.
    May cause respiratory irritation.
    Combustible liquid.
    Causes skin irritation.

    GHS P Statement
    P210-P261-P280-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit