Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytiva Phenyl Sepharose™ 6 Fast Flow Chromatography Media

Catalog No. 45002937 Shop All Cytiva Products
Change view
Click to view available options
Quantity:
25 mL
200 mL
Product Type:
Phenyl Sepharose 6 Fast Flow Media (high sub)
Phenyl Sepharose 6 Fast Flow Media (low sub)
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Product Type
45-002-937 200 mL Phenyl Sepharose 6 Fast Flow Media (low sub)
45-002-939 200 mL Phenyl Sepharose 6 Fast Flow Media (high sub)
45-002-938 25 mL Phenyl Sepharose 6 Fast Flow Media (low sub)
45-002-940 25 mL Phenyl Sepharose 6 Fast Flow Media (high sub)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 45-002-937 Supplier Cytiva Supplier No. 17096505
Add to Cart
Add to Cart

Used for capture and intermediate purification requiring low to medium hydrophobicity

  • Base matrix derivatized via uncharged, chemically-stable ether linkages
  • Two levels of substitution help find optimal selectivity and binding capacity for given application
  • Hydrophilic nature of the base matrix ensures low levels of non-specific binding
  • Hydrophobic interaction chromatography (HIC) media composed of cross-linked 6% agarose beads modified with standard aromatic phenyl groups via uncharged, chemically-stable ether linkages
  • Available in range of different bulk pack sizes and convenient pre-packed formats for easy scale-up and process development
  • Degree of substitution for low sub HIC media is ∽25μmol phenyl/mL gel
  • Degree of substitution for high sub HIC media is ∽40μmol phenyl/mL gel

TRUSTED_SUSTAINABILITY

Specifications

Quantity 200 mL
For Use With (Application) For Capture and Intermediate PurifiCatIon Requiring Low to Medium Hydrophobicity
Product Type Phenyl Sepharose 6 Fast Flow Media (low sub)
Flow Rate 200 to 400 cm/hr
Matrix 6% Cross-Linked Agarose
pH Range 3 to 13
Ligand Type Phenyl
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.