Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phenyl Stearate 98.0+%, TCI America™
SDP

Catalog No. O000725G
Click to view available options
Quantity:
5 g
25 g

Chemical Identifiers

CAS 637-55-8
Molecular Formula C24H40O2
Molecular Weight (g/mol) 360.58
MDL Number MFCD00026663
InChI Key NUMNZKICGJJSHN-UHFFFAOYSA-N
Synonym phenyl stearate, octadecanoic acid, phenyl ester, stearic acid phenyl ester, stearic acid, phenyl ester, phenylstearate, acmc-209nhu, octadecanoicacid,phenylester, stearic acid, phenyl ester 8ci
PubChem CID 69477
IUPAC Name phenyl octadecanoate
SMILES CCCCCCCCCCCCCCCCCC(=O)OC1=CC=CC=C1

Specifications

Melting Point 53°C
Color White
Boiling Point 267°C
Quantity 25 g
Formula Weight 360.58
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Phenyl Stearate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.