Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phenyl Sulfide 99%, ACROS Organics™

Click to view available options
Quantity:
25mL
100mL
Packaging:
Glass bottle
139-66-2
C12H10S
186.27
MFCD00003064
LTYMSROWYAPPGB-UHFFFAOYSA-N
diphenyl sulfide,phenyl sulfide,phenylthiobenzene,diphenyl sulphide,diphenyl thioether,benzene, 1,1'-thiobis,diphenylmercaptan,diphenylthiamethane,diphenyl monosulfide,sulfide, diphenyl
8766
CHEBI:38959
phenylsulfanylbenzene
C1=CC=C(C=C1)SC2=CC=CC=C2
139-66-2
C12H10S
186.27
MFCD00003064
LTYMSROWYAPPGB-UHFFFAOYSA-N
diphenyl sulfide,phenyl sulfide,phenylthiobenzene,diphenyl sulphide,diphenyl thioether,benzene, 1,1'-thiobis,diphenylmercaptan,diphenylthiamethane,diphenyl monosulfide,sulfide, diphenyl
8766
CHEBI:38959
phenylsulfanylbenzene
C1=CC=C(C=C1)SC2=CC=CC=C2
Boiling Point 296°C
Melting Point -40°C
Flash Point >112°C
Specific Gravity 1.11
Refractive Index 1.6317 to 1.6337
Density 1.1100g/mL
Quantity 100mL
Infrared Spectrum Authentic
Color Colorless to Yellow
Packaging Glass bottle
Assay Percent Range 99%
Linear Formula (C6H5)2S
Beilstein 06, 299
Fieser 06,241
Merck Index 15, 3371
Solubility Information Solubility in water: insoluble in water. Other solubilities: soluble in hot alcohol,miscible with benzene,ether and carbon disulfide
Formula Weight 186.27
Physical Form Liquid
Percent Purity ≥98.5%
Chemical Name or Material Phenyl sulfide, 99%
Show More Show Less

GHS H Statement:
Causes serious eye irritation.
Harmful if swallowed.
Causes skin irritation.

GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

STENCH

EINECSNumber : 205-371-4

RTECSNumber : SX2275000

TSCA : TSCA