Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phenyl sulfoxide, 97%

Catalog No. p-184128
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle

CAS: 945-51-7 | C12H10OS | 202.27 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 945-51-7
Molecular Formula C12H10OS
Molecular Weight (g/mol) 202.27
MDL Number MFCD00002085
InChI Key JJHHIJFTHRNPIK-UHFFFAOYSA-N
Synonym diphenyl sulfoxide, phenyl sulfoxide, sulfinyldibenzene, phenylsulfinylbenzene, diphenyl sulphoxide, sulfoxide, diphenyl, diphenylsulfoxide, benzene, 1,1'-sulfinylbis, 1,1'-sulfinyldibenzene, phenylsulfinyl benzene
PubChem CID 13679
IUPAC Name benzenesulfinylbenzene
SMILES C1=CC=C(C=C1)S(=O)C2=CC=CC=C2

Specifications

Melting Point 66.0°C to 71.0°C
Color White
Boiling Point 206.0°C to 208.0°C (13.0 mmHg)
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass bottle
Linear Formula (C6H5)2SO
Quantity 25 g
Beilstein 06, 300
Fieser 01,348; 17,365
Formula Weight 202.27
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material Phenyl sulfoxide, 97%
Show More Show Less
EINECSNumber 213-415-9
RTECSNumber DA9185000
TSCA TSCA

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.