Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phenyl vinyl sulfide, 97%, ACROS Organics™

Click to view available options
Quantity:
1mL
Packaging:
Glass bottle
1822-73-7
C8H8S
136.21
MFCD00009646
GMPDOIGGGXSAPL-UHFFFAOYSA-N
phenyl vinyl sulfide, phenylthioethene, phenyl vinyl sulphide, phenylthio ethylene, vinylthiobenzene, benzene, ethenylthio, vinyl phenyl sulfide, sulfide, phenyl vinyl, vinylsulfanyl benzene, phenyl vinyl thioether
74572
ethenylsulfanylbenzene
C=CSC1=CC=CC=C1
1822-73-7
C8H8S
136.21
MFCD00009646
GMPDOIGGGXSAPL-UHFFFAOYSA-N
phenyl vinyl sulfide, phenylthioethene, phenyl vinyl sulphide, phenylthio ethylene, vinylthiobenzene, benzene, ethenylthio, vinyl phenyl sulfide, sulfide, phenyl vinyl, vinylsulfanyl benzene, phenyl vinyl thioether
74572
ethenylsulfanylbenzene
C=CSC1=CC=CC=C1
Boiling Point 94°C to 95°C (25.0mmHg)
Flash Point 45°C
Refractive Index 1.5877 to 1.5897
Specific Gravity 1.04
Density 1.0400g/mL
Packaging Glass bottle
Quantity 1mL
Assay Percent Range 97%
Linear Formula C6H5SCH=CH2
Beilstein 06, II, 288
Formula Weight 136.21
Percent Purity 97%
Chemical Name or Material Phenyl vinyl sulfide, 97%
Show More Show Less

GHS H Statement:
Flammable liquid and vapor.

GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 217-351-2