Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cobalt(II) sulfate heptahydrate, 99+%, extra pure
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Glass bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Suitable for battery materials development.

Specifications
Specifications
Melting Point | 98.0°C |
Color | Red to Brown |
Physical Form | Crystalline Powder and/or Crystals |
Quantity | 500 g |
Assay Percent Range | 99% min. (Complexometry) |
Linear Formula | CoSO4·7H2O |
Merck Index | 15,2432 |
Solubility Information | Solubility in water: 362g/L (20°C). Other solubilities: slightly soluble in methanol and ethanol |
IUPAC Name | cobalt(2+);sulfate;heptahydrate |
Formula Weight | 281.09 |
Show More |
Safety and Handling
Hazard Category | GHS Signal Word: Danger |
Hazard Statement | GHS H Statement May cause an allergic skin reaction. Very toxic to aquatic life with long lasting effects. May cause allergy or asthma symptoms or breathing difficulties if inhaled. Suspected of causing genetic defects. May |
Precautionary Statement | GHS P Statement Avoid breathing dust/fume/gas/mist/vapors/spray. IF INHALED: If breathing is difficult,remove to fresh air and keep at rest in a position comfortable for breathing. If experiencing respiratory symptoms: Call a POISON CENT |
RTECSNumber | GG3200000 |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction