Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phenyl Vinyl Sulfone 98.0+%, TCI America™
SDP

Catalog No. P098225G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P098225G 25 g
P09825G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P098225G Supplier TCI America Supplier No. P098225G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 5535-48-8
Molecular Formula C8H8O2S
Molecular Weight (g/mol) 168.21
MDL Number MFCD00007554
InChI Key UJTPZISIAWDGFF-UHFFFAOYSA-N
Synonym phenyl vinyl sulfone, vinylsulfonyl benzene, ethenesulfonyl benzene, benzene, ethenylsulfonyl, sulfone, phenyl vinyl, vinylsulfonylbenzene, phenyl vinyl sulphone, ethenylsulfonyl benzene, ethenesulfonyl-benzene, phenyl-vinylsulfon
PubChem CID 79664
IUPAC Name (ethenesulfonyl)benzene
SMILES C=CS(=O)(=O)C1=CC=CC=C1

Specifications

Melting Point 68°C
Color White-Yellow
Quantity 25 g
Formula Weight 168.21
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Phenyl Vinyl Sulfone
EINECSNumber (3)-4398
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.