Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phenylacetyl chloride, 98%

Catalog No. AAB2498736
Click to view available options
Quantity:
100 g
500 g

CAS: 103-80-0 | C8H7ClO | 154.59 g/mol

Phenylacetyl chloride is used in the preparation of 1,2-diphenyl-ethanone by reaction with iodobenzene using tetrabutylammonium tetrafluoroborate as a phase transfer catalyst. It is widely used in the chemical industry for manufacturing dyes and pharmaceutical products. It plays an important role in the preparation of penicillin. Further, it is used to prepare esters for flavorings.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Phenylacetyl chloride is used in the preparation of 1,2-diphenyl-ethanone by reaction with iodobenzene using tetrabutylammonium tetrafluoroborate as a phase transfer catalyst. It is widely used in the chemical industry for manufacturing dyes and pharmaceutical products. It plays an important role in the preparation of penicillin. Further, it is used to prepare esters for flavorings.

Solubility
Miscible with alcohol and ether.

Notes
Moisture sensitive. Store in a cool place.Incompatible with amines, most common metals, strong bases and strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 103-80-0
Molecular Formula C8H7ClO
Molecular Weight (g/mol) 154.59
MDL Number MFCD00000729
InChI Key VMZCDNSFRSVYKQ-UHFFFAOYSA-N
Synonym phenylacetyl chloride, benzeneacetyl chloride, acetyl chloride, phenyl, phenylacetic acid chloride, phenacetyl chloride, phenylacetylchloride, unii-t30899drnd, phenyl acetyl chloride, a-toluoyl chloride, phenylacetyl
PubChem CID 7679
IUPAC Name 2-phenylacetyl chloride
SMILES ClC(=O)CC1=CC=CC=C1

Specifications

Density 1.168
Boiling Point 225°C
Flash Point 102°C (215°F)
Refractive Index 1.5325
Quantity 500 g
UN Number UN2577
Beilstein 742254
Sensitivity Moisture sensitive
Solubility Information Miscible with alcohol and ether.
Formula Weight 154.6
Percent Purity 98%
Chemical Name or Material Phenylacetyl chloride
Show More Show Less
Hazard Category H314-H500
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: PHENYLACETYL CHLORIDE
EINECSNumber 203-146-5
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.