Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phenylacetylene, 98%, pure

Catalog No. AC152465000
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle

CAS: 536-74-3 | C8H6 | 102.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 536-74-3
Molecular Formula C8H6
Molecular Weight (g/mol) 102.14
InChI Key UEXCJVNBTNXOEH-UHFFFAOYSA-N
Synonym phenylacetylene, phenylethyne, benzene, ethynyl, 1-phenylethyne, phenylacetylide, phenyl acetylene, ethyne, phenyl, acetylene, phenyl, ethynyl-benzene, phenylacethylene
PubChem CID 10821
IUPAC Name ethynylbenzene
SMILES C#CC1=CC=CC=C1

Specifications

Melting Point -44.8°C
Color Colorless to Yellow
Density 0.9300g/mL
Boiling Point 142.0°C to 144.0°C
Flash Point 31°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.5470 to 1.5500
Linear Formula C6H5C≡CH
Quantity 500 g
Beilstein 05,511
Merck Index 15,391
Specific Gravity 0.93
Solubility Information Solubility in water: insoluble. Other solubilities: miscible with alcohol,ether and other organic,solvents
Formula Weight 102.14
Percent Purity 98%
Grade Pure
Physical Form Liquid
Chemical Name or Material Phenylacetylene
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Flammable liquid and vapor.
Precautionary Statement GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
EINECSNumber 208-645-1

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.