Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phenylarsine Oxide, 0.025 N, AquaPhoenix Scientific™
SCI_ED

Catalog No. S88701 Shop All AquaPhoenix Scientific Products
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S88701 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S88701 Supplier Aqua Phoenix Scientific Supplier No. PO4410P
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Standardized titrant for laboratory analytical applications.

Chemical Identifiers

CAS 1310-73-2 , 637-03-6 , 7732-18-5
Molecular Formula HNaO
Molecular Weight (g/mol) 40.00
InChI Key HEMHJVSKTPXQMS-UHFFFAOYSA-M
IUPAC Name sodium hydroxide
SMILES [OH-].[Na+]

Specifications

Quantity 500 mL
Chemical Name or Material Phenylarsine Oxide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.