Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Phenylarsine Oxide (PAO) Titrant, 0.00500 N (N/200), Ricca Chemical
Shop All Ricca Chemical Company Products

Click to view available options
Quantity:
500 mL
1 L
4 L
Packaging:
Amber Glass Bottle
Specifications
Specifications
Color | Colorless to Tan |
CAS Min % | 0.05, 98.0, 0.02 |
CAS Max % | 0.05, 100.0, 0.02 |
Grade | Laboratory |
Packaging | Amber Glass Bottle |
Treatment(s) | Stabilized |
pH | 6 to 8.5 |
Quantity | 4 L |
Physical Form | Liquid |
Concentration or Composition Notes | 0.00500Normal |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction