Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phenylarsonic acid, 97%

Catalog No. p-7080893
Click to view available options
Quantity:
100 g

CAS: 98-05-5 | C6H7AsO3 | 202.041 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 98-05-5
Molecular Formula C6H7AsO3
Molecular Weight (g/mol) 202.041
MDL Number MFCD00002097
InChI Key LVKZSFMYNWRPJX-UHFFFAOYSA-N
Synonym benzenearsonic acid, monophenylarsonic acid, arsonic acid, phenyl, unii-57f9ku116m, kyselina benzenarsonova czech, arsonic acid, as-phenyl, kyselina benzenarsonova, benzenarsonic acid, benzenearsonie acid, acmc-20ajwq
PubChem CID 7365
ChEBI CHEBI:29851
IUPAC Name phenylarsonic acid
SMILES C1=CC=C(C=C1)[As](=O)(O)O

Specifications

Melting Point 158°C to 162°C (decomposition)
Quantity 100 g
UN Number UN3465
Merck Index 14,1067
Formula Weight 202.04
Percent Purity 97%
Chemical Name or Material Phenylarsonic acid
Hazard Category H301+H331
Hazard Statement GHS H Statement
H301-H331
Toxic if swallowed.
Toxic if inhaled.
Precautionary Statement P261-P264b-P270-P271-P301+P310-P304+P340-P311-P330-P501c
DOTInformation Hazard Class: 6.1; Packaging Group: I
EINECSNumber 202-631-9
RTECSNumber CY3150000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.