Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phenylazoresorcinol 80.0+%, TCI America™
SDP

Catalog No. P01511G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P01511G 1 g
P015125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P01511G Supplier TCI America Supplier No. P01511G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 67503-46-2
Molecular Formula C18H14N2O4
Molecular Weight (g/mol) 322.32
MDL Number MFCD00059881
InChI Key KVQRGKQWGHNYJE-FMQUCBEESA-N
PubChem CID 77519172
IUPAC Name 3-[(E)-(3-hydroxyphenoxy)diazenyl]oxy-2-phenylphenol
SMILES C1=CC=C(C=C1)C2=C(C=CC=C2ON=NOC3=CC=CC(=C3)O)O

Specifications

Quantity 1 g
Formula Weight 214.22
Percent Purity ≥80.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Phenylazoresorcinol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.