Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phenylboronic acid, 97%, ACROS Organics™

Click to view available options
Quantity:
10g
50g
250g
Packaging:
Glass bottle
98-80-6
C6H7BO2
121.93
HXITXNWTGFUOAU-UHFFFAOYSA-N
benzeneboronic acid,boronic acid, phenyl,phenyl boronic acid,phenylboric acid,dihydroxyphenylborane,phenyldihydroxyborane,dihydroxy phenyl borane,phenylboronicacid,borophenylic acid,usaf bo-2
66827
CHEBI:44923
phenylboronic acid
B(C1=CC=CC=C1)(O)O
98-80-6
C6H7BO2
121.93
HXITXNWTGFUOAU-UHFFFAOYSA-N
benzeneboronic acid,boronic acid, phenyl,phenyl boronic acid,phenylboric acid,dihydroxyphenylborane,phenyldihydroxyborane,dihydroxy phenyl borane,phenylboronicacid,borophenylic acid,usaf bo-2
66827
CHEBI:44923
phenylboronic acid
B(C1=CC=CC=C1)(O)O
Infrared Spectrum Authentic
Packaging Glass bottle
Color White
Melting Point 217°C to 222°C
Quantity 10g
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 98+%
Linear Formula C6H5B(OH)2
Beilstein 16, 920
Fieser 01,833; 02,317; 03,221; 05,513; 07,284; 09,23; 11,408; 15,254
Merck Index 15, 1070
Solubility Information Solubility in water: 10g/L (20°C). Other solubilities: solubility in benzene : 1.75%, xylene : 1.2 %, methanol : 178 %, ether : 30.2%
Formula Weight 121.93
Physical Form Crystalline Powder
Percent Purity ≥98.0%
Chemical Name or Material Phenylboronic acid
Show More Show Less

GHS H Statement:
Harmful if swallowed.
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Warning