Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phenylethyl Alcohol Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™

Catalog No. p-7187723 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
2 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-4828 2 g
11-101-4827 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 11-101-4828 Supplier MilliporeSigma Supelco Supplier No. PHR11222G
Only null left
Add to Cart
Add to Cart
Cleaning Products, Cosmetics, Food and Beverages, Personal Care and Pharmaceutical (small molecule)
TRUSTED_SUSTAINABILITY

Specifications

CAS 60-12-8
Vapor Pressure 1 mmHg (58°C)
Vapor Density 4.21 (vs. air)
Density 1.020 g/mL (at 20°C)
Flash Point 102°C - Closed Cup
Physical Form Neat
Linear Formula C6H5 CH2 CH2 OH
MDL Number MFCD00002886
Melting Point -27°C
Boiling Point 219°C to 221°C (750 mmHg)
Synonym 2-Phenylethanol; beta-PEA; 2-Phenylethyl alcohol; Benzyl carbinol; PEA
Type Certified Reference Material
Quantity 2 g
Formula Weight 122.16
Grade Certified Reference Material
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.