Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phenylphosphinic acid, 99%, Thermo Scientific™

Catalog No. AC130780050
Click to view available options
:
5g; Glass bottle

Specifications

CAS 1779-48-2
Melting Point 82.0°C to 87.0°C
Color White
Boiling Point 180.0°C
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Molecular Formula C6H7O2P
Linear Formula C6H5P(O)(OH)H
Quantity 5 g
MDL Number MFCD00002131
Beilstein 16, 791
Synonym phenylphosphinic acid, benzenephosphinic acid, phosphinic acid, phenyl, benzenephosphonous acid, phenyl phosphinic acid, mlchbqkmvknbov-uhfffaoysa-n, benzolphosphonigs, aure, benzene phosphinic acid, hydroxyphenylphosphineoxide, acmc-1bt5t
Solubility Information Solubility in water: 7.7g/100mL water (25°C). Other solubilities: soluble in alcohol,acetone,slightly soluble in ether,insoluble in benzene,hexane,ccl4
InChI Key MLCHBQKMVKNBOV-UHFFFAOYSA-N
SMILES OP(=O)C1=CC=CC=C1
IUPAC Name hydroxy-oxo-phenylphosphanium
Molecular Weight (g/mol) 142.09
PubChem CID 6327116
Formula Weight 142.09
Percent Purity 99%
Physical Form Crystalline Solid
Chemical Name or Material Phenylphosphinic acid, 99%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Causes serious eye damage.
May cause respiratory irritation.
Causes severe skin burns and eye damage.
Harmful if swallowed.
Precautionary Statement GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 217-217-3
RTECSNumber SZ5462500
TSCA TSCA

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.