Learn More
Phenylphosphonic acid, 98%, Thermo Scientific Chemicals
Description
Phenylphosphonic acid additive used with Noyori's catalyst for oxidation of sulfides to sulfones. Used as additive of unsaturated polyester PU resin. It also can be added into nylon to improve its polymerization degree. It can be used as extreme pressure agent of lube; used as catalyst of carboxylic acid & alcohol reaction; it can be used as fire retarding treatment of fiber used asganic materials.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Phenylphosphonic acid additive used with Noyori′s catalyst for oxidation of sulfides to sulfones. Used as additive of unsaturated polyester PU resin. It also can be added into nylon to improve its polymerization degree. It can be used as extreme pressure agent of lube; used as catalyst of carboxylic acid & alcohol reaction; it can be used as fire retarding treatment of fiber used asganic materials.
Solubility
Soluble in water.
Notes
Store in cool, dry place in a well sealed container. Store away from oxidizing agents.

Specifications
Specifications
Melting Point | 160°C to 162°C |
Quantity | 100 g |
UN Number | UN3261 |
Beilstein | 2245168 |
Solubility Information | Soluble in water. |
Formula Weight | 158.09 |
Percent Purity | 98% |
Chemical Name or Material | Phenylphosphonic acid |
Safety and Handling
Hazard Category | H302-H315-H318-H335 |
Hazard Statement | GHS H Statement H314-H318-H302 Causes severe skin burns and eye damage. Causes serious eye damage. Harmful if swallowed. |
Precautionary Statement | P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P310-P312-P330-P332+P313-P362-P501c |
DOTInformation | Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S. |
EINECSNumber | 216-388-1 |
RTECSNumber | TA0350000 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.