Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phenylphosphonic Acid 98.0+%, TCI America™
SDP

Catalog No. P0204100G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P0204100G 100 g
P020425G 25 g
P0204500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. P0204100G Supplier TCI America Supplier No. P0204100G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 1571-33-1
Molecular Formula C6H7O3P
Molecular Weight (g/mol) 158.09
MDL Number MFCD00002136
InChI Key QLZHNIAADXEJJP-UHFFFAOYSA-N
Synonym benzenephosphonic acid, phosphonic acid, phenyl, phenyl-phosphonic acid, phosphonic acid, p-phenyl, phenyl phosphonic acid, unii-byd76t2868, sv7, pubchem21303, phosphonic acid,phenyl, wln: qpqo&r
PubChem CID 15295
IUPAC Name phenylphosphonic acid
SMILES OP(O)(=O)C1=CC=CC=C1

Specifications

Melting Point 163°C
Color White
Quantity 100 g
Formula Weight 158.09
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Phenylphosphonic Acid
EINECSNumber (3)-2524
RTECSNumber TA0350000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.