Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phenylphosphonic dichloride, 97%

Catalog No. p-183663
Click to view available options
Quantity:
250 mL
1 L
Packaging:
Glass bottle

CAS: 824-72-6 | C6H5Cl2OP | 194.98 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 824-72-6
Molecular Formula C6H5Cl2OP
Molecular Weight (g/mol) 194.98
MDL Number MFCD00002070
InChI Key IBDMRHDXAQZJAP-UHFFFAOYSA-N
Synonym phenylphosphonic dichloride, phosphonic dichloride, phenyl, benzenephosphonic dichloride, phenylphosphonyl dichloride, dichlorophenylphosphine oxide, phenyldichlorophosphine oxide, benzenephosphonyl chloride, phenylphosphonic acid dichloride, phenylphosphonodichloridic acid, phosphonic dichloride, p-phenyl
PubChem CID 69990
IUPAC Name dichlorophosphorylbenzene
SMILES ClP(Cl)(=O)C1=CC=CC=C1

Specifications

Melting Point 3.0°C
Color Yellow to Brown
Density 1.3900g/mL
Boiling Point 258.0°C
Flash Point 204°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (Argentometry)
Packaging Glass bottle
Refractive Index 1.5570 to 1.5610
Linear Formula C6H5P(O)Cl2
Quantity 250 mL
Beilstein 16, 804
Fieser 01,846; 16,284
Specific Gravity 1.39
Solubility Information Solubility in water: reacts
Formula Weight 194.98
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material Phenylphosphonic dichloride, 97%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes severe skin burns and eye damage.
Harmful if swallowed.
May be corrosive to metals.
Reacts violently with water.
Precautionary Statement GHS P Statement
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre
EINECSNumber 212-534-3
TSCA TSCA

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.