Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phenylsilane, 97%

Catalog No. p-7023892
Click to view available options
Quantity:
5 g
25 g
100 g

Catalog No. AAA1790106

Supplier: Thermo Scientific Chemicals A1790106

Only null left
Add to Cart

CAS: 694-53-1 | C6 H8 Si | 108.22 g/mol

It is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as a pharmaceutical intermediate.

Solubility
Contact with water releases a flammable alcohol. Soluble in water (Contact with water releases flammable gases), and most common organic solvents.

Notes
Moisture Sensitive. Store away from water/moisture. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Protect against electrostatic charges. Keep ignition sources away. Store under inert gas.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 694-53-1
Molecular Formula C6 H8 Si
Molecular Weight (g/mol) 108.22
MDL Number MFCD00013478
InChI Key XJWOWXZSFTXJEX-UHFFFAOYSA-N
Synonym phenylsilane, benzene, silyl, silylbenzene, silane, phenyl, fenylsilan, fenylsilan czech, phenylsilane, silyl, silane, phenyl-,
PubChem CID 6327628
IUPAC Name phenylsilicon
SMILES C1=CC=C(C=C1)[Si]

Specifications

Boiling Point 120°C
Quantity 5 g
UN Number UN3399
Flash Point 7°C (45°F)
Solubility Information Contact with water releases a flammable alcohol. Soluble in water (Contact with water releases flammable gases),and most common organic solvents.
Formula Weight 108.22
Percent Purity 97%
Refractive Index 1.5105
Sensitivity Moisture sensitive
Density 0.88
Chemical Name or Material Phenylsilane
Show More Show Less
Hazard Category H225-H261-H302+H332-H315-H319-H335
Hazard Statement GHS H Statement
H225-H261-H302-H332-H315-H319-H335
Highly flammable liquid and vapour.
In contact with water releases flammable gas.
Harmful if swallowed.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P223-P231+P232-P233-P235-P240-P241-P242-P243-P261-P264b-P270-P271-P280-P301+P312-P303+P361+P353-P304+P340-P305+P351+P338-P312-P330-P332+P313-P335+P334-P363-P370+P378q-P50
DOTInformation Transport Hazard Class: 4.3; Packing Group: II; Proper Shipping Name: ORGANOMETALLIC SUBSTANCE, LIQUID, WATER-REACTIVE, FLAMMABLE
EINECSNumber 211-772-5
TSCA Yes
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.