Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phenylsilane 97.0+%, TCI America™
SDP

Catalog No. P129125ML Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
5 mL
25 mL

Catalog No. P129125ML

Supplier: TCI America P129125ML

Only null left
Add to Cart
Add to Cart

Chemical Identifiers

CAS 694-53-1
Molecular Formula C6H5Si
Molecular Weight (g/mol) 105.191
MDL Number MFCD00013478
InChI Key XJWOWXZSFTXJEX-UHFFFAOYSA-N
Synonym phenylsilane, benzene, silyl, silylbenzene, silane, phenyl, fenylsilan, fenylsilan czech, phenylsilane, silyl, silane, phenyl-,
PubChem CID 6327628
IUPAC Name phenylsilicon
SMILES C1=CC=C(C=C1)[Si]

Specifications

Color Yellow
Boiling Point 120°C
Quantity 25 mL
UN Number 1993
Formula Weight 108.22
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material Phenylsilane
RTECSNumber VV4825000
TSCA Yes
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.