Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phenylsuccinic Anhydride 98.0+%, TCI America™
SDP

Catalog No. p-7135480
Click to view available options
Quantity:
5 g
25 g

Chemical Identifiers

CAS 1131-15-3
Molecular Formula C10H8O3
Molecular Weight (g/mol) 176.17
MDL Number MFCD00192395
InChI Key HDFKMLFDDYWABF-UHFFFAOYNA-N
Synonym phenylsuccinic anhydride, 3-phenyldihydrofuran-2,5-dione, monophenyl succinic anhydride, 2,5-furandione, dihydro-3-phenyl, acmc-20ajwd, acmc-20mmqc, phenylsuccinicanhydride, 2,5-furandione, dihydro-3-phenyl-, 3r, phenyl succinic anhydride
PubChem CID 136911
IUPAC Name 3-phenyloxolane-2,5-dione
SMILES O=C1CC(C(=O)O1)C1=CC=CC=C1

Specifications

Melting Point 54°C
Boiling Point 195°C
Quantity 5 g
Formula Weight 176.17
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material Phenylsuccinic Anhydride
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.