Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phenylsulfonylacetone, 97%

Catalog No. AAA1408503
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1408503 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAA1408503 Supplier Thermo Scientific Chemicals Supplier No. A1408503

CAS: 5000-44-2 | C9H10O3S | 198.236 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5000-44-2
Molecular Formula C9H10O3S
Molecular Weight (g/mol) 198.236
MDL Number MFCD00047794
InChI Key YBLGSNMIIPIRFC-UHFFFAOYSA-N
Synonym phenylsulfonylacetone, benzenesulfonylacetone, 1-phenylsulfonyl propan-2-one, 1-benzenesulfonyl propan-2-one, benzenesulphonylacetone, 1-phenylsulfonyl acetone, phenyl sulfonyl acetone, phenylsulphonylacetone, phenylsulfonyl acetone, 1-phenylsulfonyl-2-propanone
PubChem CID 735827
IUPAC Name 1-(benzenesulfonyl)propan-2-one
SMILES CC(=O)CS(=O)(=O)C1=CC=CC=C1

Specifications

Melting Point 54°C to 58°C
Quantity 1 g
Beilstein 640928
Formula Weight 198.24
Percent Purity 97%
Chemical Name or Material Phenylsulfonylacetone
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.