Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Phenylthiophosphonic Dichloride 98.0+%, TCI America™

Click to view available options
Quantity:
25 g
500 g
Specifications
Specifications
Color | Yellow |
Boiling Point | 270°C |
Quantity | 25 g |
UN Number | 3265 |
Formula Weight | 211.04 |
Percent Purity | ≥98.0% (GC,T) |
Physical Form | Liquid |
Chemical Name or Material | Phenylthiophosphonic Dichloride |
Safety and Handling
EINECSNumber | (3)-2511&(3)-2612 |
TSCA | Yes |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction