Learn More
Eisco™ Phenylthiourea (PTC) Paper Test Strips
SCI_ED
Catalog No. S80844 Shop All Eisco Products
Change view
Click to view available options
Quantity:
100/Pk., 12 Pk./Cs.
Catalog No. Quantity
S80844 100/Pk., 12 Pk./Cs.
1 options
Catalog No. S80844 Supplier Eisco™ Supplier No. FSC1031PK12
Add to Cart
Add to Cart
Genetic Taste Testing Phenylthiourea (PTC) Paper Strips are used to demonstrate the effects of heredity on taste
  • Only 7 out of 10 people can taste PTC
  • Perfect for a demonstration of genetics and heredity
  • 100 Strips/Vial, 12 Vials/Pk
Specifications
Product Type Test Strips, Phenylthiourea (PTC)
Description Paper Taste Test
For Use With (Application) Studying The Effects of Genetics and Heredity on Taste
Quantity 100/Pk., 12 Pk./Cs.