Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Phenyltrimethylammonium chloride, 98+%, Thermo Scientific Chemicals
Click to view available options
Quantity:
100 g
500 g
Description
Phenyltrimethylammonium chloride is an effective phase transfer catalyst, methylating agent, also useful for alkaline depolymerizations, corrosion inhibitor for low carbon steel.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Phenyltrimethylammonium chloride is an effective phase transfer catalyst, methylating agent, also useful for alkaline depolymerizations, corrosion inhibitor for low carbon steel.
Solubility
Soluble in water 333 g/L.
Notes
Hygroscopic. Store under inert gas. Store away from oxidizing agent, water/ moisture.
Phenyltrimethylammonium chloride is an effective phase transfer catalyst, methylating agent, also useful for alkaline depolymerizations, corrosion inhibitor for low carbon steel.
Solubility
Soluble in water 333 g/L.
Notes
Hygroscopic. Store under inert gas. Store away from oxidizing agent, water/ moisture.
Specifications
Melting Point | ∼236°C (sublimation) |
Quantity | 500 g |
UN Number | UN2811 |
Beilstein | 3572527 |
Sensitivity | Hygroscopic |
Solubility Information | Soluble in water 333g/L. |
IUPAC Name | trimethyl(phenyl)azanium;chloride |
Formula Weight | 171.67 |
Percent Purity | ≥98% |
Chemical Name or Material | Phenyltrimethylammonium chloride |
Safety and Handling
Hazard Category | H301+H311 |
Hazard Statement | GHS H Statement H301-H311 Toxic if swallowed. Toxic in contact with skin. |
Precautionary Statement | P264b-P270-P280-P301+P310-P302+P352-P312-P330-P361-P363-P501c |
DOTInformation | Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S. |
EINECSNumber | 205-319-0 |
RTECSNumber | BT2190000 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction