Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phenyltrimethylammonium iodide, 99%

Catalog No. aaa1440222
Click to view available options
Quantity:
100 g
500 g

CAS: 98-04-4 | C9H14IN | 263.122 g/mol

Phenyltrimethylammonium iodide is an important raw material and intermediate used in organic synthesis, pharmaceuticals, dyes and agrochemicals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Phenyltrimethylammonium iodide is an important raw material and intermediate used in organic synthesis, pharmaceuticals, dyes and agrochemicals.

Solubility
Soluble in water and methanol.

Notes
Store at room temperature. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Light sensitive. It is hygroscopic in nature. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 98-04-4
Molecular Formula C9H14IN
Molecular Weight (g/mol) 263.122
MDL Number MFCD00011791
InChI Key KKLAORVGAKUOPZ-UHFFFAOYSA-M
Synonym phenyltrimethylammonium iodide, trimethylphenylammonium iodide, n,n,n-trimethylanilinium iodide, trimethylanilinium iodide, trimethylanillinium iodide, n,n-dimethylaniline methiodide, unii-4o888xq7fp, ammonium, phenyltrimethyl-, iodide, n,n,n-trimethylbenzenaminium iodide, benzenaminium, n,n,n-trimethyl-, iodide
PubChem CID 94135
IUPAC Name trimethyl(phenyl)azanium;iodide
SMILES C[N+](C)(C)C1=CC=CC=C1.[I-]

Specifications

Melting Point ∼227°C (sublimation)
Quantity 100 g
UN Number UN2811
Beilstein 3916752
Sensitivity Light sensitive
Merck Index 14,7318
Solubility Information Soluble in water and methanol.
Formula Weight 263.12
Percent Purity 99%
Chemical Name or Material Phenyltrimethylammonium iodide
Hazard Category H315-H318-H335
Hazard Statement GHS H Statement
H311-H302-H315-H319-H335
Toxic in contact with skin.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P310-P312-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
EINECSNumber 202-630-3
RTECSNumber BT2450000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.