Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phenylundecanoic Acid, Mixture of Isomers 99%, Thermo Scientific™

Catalog No. AC417861000
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC417861000 100 g
AC417865000 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC417861000 Supplier Thermo Scientific Chemicals Supplier No. 417861000

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3343-24-6
Molecular Formula C17H26O2
Molecular Weight (g/mol) 262.393
InChI Key AUQIXRHHSITZFM-UHFFFAOYSA-N
Synonym benzeneundecanoic acid, phenylundecanoic acid, undecanoic acid, phenyl, benzeneundecanoicacid, 11-phenylundecanoicacid, #
PubChem CID 76863
IUPAC Name 11-phenylundecanoic acid
SMILES C1=CC=C(C=C1)CCCCCCCCCCC(=O)O

Specifications

Color Yellow
Infrared Spectrum Authentic
Refractive Index 1.4980 to 1.5010 (20°C, 589nm)
Linear Formula C6H5CH2(CH2)9CO2H
Quantity 100 g
Formula Weight 262.38
Percent Purity ≥98.5%
Physical Form Viscous Liquid
Chemical Name or Material Phenylundecanoic acid, mixture of isomers, 99%
EINECSNumber 222-087-6
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.