Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PHEX Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5116248
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5116248 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5116248 Supplier Invitrogen™ Supplier No. PA5116248
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

PHEX Polyclonal Antibody for Western Blot

Peptidase that cleaves SIBLING (small integrin-binding ligand, N-linked glycoprotein)-derived ASARM peptides, thus regulating their biological activity. Cleaves ASARM peptides between Ser and Glu or Asp residues. Regulates osteogenic cell differentiation and bone mineralization through the cleavage of the MEPE-derived ASARM peptide. Promotes dentin mineralization and renal phosphate reabsorption by cleaving DMP1- and MEPE-derived ASARM peptides. Inhibits the cleavage of MEPE by CTSB/cathepsin B thus preventing MEPE degradation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PHEX
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene PHEX
Gene Accession No. P78562
Gene Alias Gy; HPDR; HPDR1; HYP; HYP1; LXHR; metalloendopeptidase homolog PEX; Metalloendopeptidase homolog PEX (Phosphate regulating neutral endopeptidase) (X-linked hypophosphatemia protein) (HYP) (Vitamin D-resistant hypophosphatemic rickets protein); PEX; PHEX; phosphate regulating endopeptidase homolog, X-linked; phosphate regulating gene with homologies to endopeptidases on the X chromosome (hypophosphatemia, vitamin D resistant rickets); phosphate regulating neutral endopeptidase; Phosphate regulating neutral endopeptidase on the X chromosome (X-linked hypophosphatemia XLH); phosphate-regulating neutral endopeptidase; Phosphate-regulating neutral endopeptidase PHEX; Vitamin D-resistant hypophosphatemic rickets protein; XLH; X-linked hypophosphatemia protein; X-linked phosphate regulating endopeptidase
Gene Symbols PHEX
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 567-595 amino acids from the C-terminal region of human PHEX.
Purification Method Antigen affinity chromatography, Protein A
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5251
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.