Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PHF1 Mouse anti-Human, Clone: 2G7, Abnova™
Mouse Monoclonal Antibody
Supplier: Abnova Corporation H00005252M04A
Description
Mouse monoclonal antibody raised against a partial recombinant PHF1.
- High specificity
- High reactivity
Specifications
PHF1 | |
Monoclonal | |
Unconjugated | |
ascites with no preservative | |
NM_024165 | |
PHF1 | |
PHF1 (NP_077084, 2 a.a. ∼ 100 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
RUO | |
5252 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
Ascites |
ELISA, Western Blot | |
2G7 | |
Mouse monoclonal antibody raised against a partial recombinant PHF1. | |
PHF1 | |
MTF2L2/PCL1/PHF2 | |
Mouse | |
200 μL | |
Primary | |
Human | |
Antibody | |
IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction