Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PHF1 Mouse anti-Mouse, Human, Clone: 12C2.1, MilliporeSigma™

Catalog No. MABE676MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABE676MI 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABE676MI Supplier MilliporeSigma Supplier No. MABE676
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-PHF1, clone 12C2.1 is a highly specific mouse monoclonal antibody that targets PHF1 and has been tested in Immunohistochemistry (Paraffin) and Western Blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PHF1
Applications Western Blot, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 12C2.1
Concentration Please refer to lot specific datasheet.
Conjugate Unconjugated
Gene Symbols PHF1,PCL1
Host Species Mouse
Immunogen GST/His-tagged recombinat fragment corresponding to 137 amino acids from human PHD finger protein 1.
Quantity 100 μL
Regulatory Status RUO
Research Discipline Epigenetics and Nuclear Function
Primary or Secondary Primary
Gene ID (Entrez) NP_002627
Test Specificity Clone 12C2.1 specifically detects PHF1 in human tissues. It targets an epitope within 137 amino acids from the internal region.
Target Species Mouse, Human
Content And Storage Stable for 1 year at 2-8°C from date of receipt.
Form Purified
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.