Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ PHF1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5101061
Description
Antibody detects endogenous levels of total PHF1.
This gene encodes a protein with significant sequence similarity to Drosophila Polycomblike. The encoded protein contains a zinc finger-like PHD (plant homeodomain) finger which is distinct from other classes of zinc finger motifs and which shows the typical Cys4-His-Cys3 arrangement. PHD finger genes are thought to belong to a diverse group of transcriptional regulators possibly affecting eukaryotic gene expression by influencing chromatin structure. Two transcript variants have been found for this gene.
Specifications
PHF1 | |
Polyclonal | |
Unconjugated | |
PHF1 | |
AW557215; D17Ertd455e; hPCl1; hPHF1; mPCl1; MTF2L2; PCL1; PHD finger protein 1; PHF1; Phf2; Plc1; polycomb-like 1; polycomb-like protein 1; Protein PHF1; T-complex testis-expressed 3; Tctex3; Tctex-3; TDRD19C; testicular tissue protein Li 140; tudor domain containing 19C | |
Rabbit | |
Affinity chromatography | |
RUO | |
21652, 294287, 5252 | |
-20°C | |
Liquid |
Western Blot, Immunocytochemistry | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
O43189, Q9Z1B8 | |
PHF1 | |
A synthesized peptide derived from human PHF1(Accession O43189), corresponding to amino acid residues V361-A411. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction