Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ PHF1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA556621
Description
Immunogen sequence: ACTQCLSKPL LYGDRFYEFE CCVCRGGPEK VRRLQLRWVD VAHLVLYHLS VCCKKKYFDF DREILPFTSE NWDSLLLGEL SDTPKGERSS KLLSALNSHK DRFISGREIK KRKCLFGLHA RMPPPVEPPT GDGALTS Highest antigen sequence identity to the following orthologs: Mouse - 96%, Rat - 96%.
This gene encodes a protein with significant sequence similarity to Drosophila Polycomblike. The encoded protein contains a zinc finger-like PHD (plant homeodomain) finger which is distinct from other classes of zinc finger motifs and which shows the typical Cys4-His-Cys3 arrangement. PHD finger genes are thought to belong to a diverse group of transcriptional regulators possibly affecting eukaryotic gene expression by influencing chromatin structure. Two transcript variants have been found for this gene.
Specifications
PHF1 | |
Polyclonal | |
Unconjugated | |
PHF1 | |
AW557215; D17Ertd455e; hPCl1; hPHF1; mPCl1; MTF2L2; PCL1; PHD finger protein 1; PHF1; Phf2; Plc1; polycomb-like 1; polycomb-like protein 1; Protein PHF1; T-complex testis-expressed 3; Tctex3; Tctex-3; TDRD19C; testicular tissue protein Li 140; tudor domain containing 19C | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
5252 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
O43189 | |
PHF1 | |
Recombinant protein corresponding to Human PHF1. Recombinant protein control fragment (Product #a href=. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction