Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PHF1 Rabbit anti-Human, Polyclonal, MilliporeSigma™

Catalog No. ABE86325UL Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
25 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ABE86325UL 25 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ABE86325UL Supplier MilliporeSigma Supplier No. ABE86325UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Anti-PHF1 is a highly specific rabbit polyclonal antibody that targets PHD finger protein 1 and has been tested in Immunohistochemistry (Paraffin) and Western Blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PHF1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration Please refer to lot specific datasheet.
Conjugate Unconjugated
Gene Symbols PHF1,PCL1
Host Species Rabbit
Immunogen GST-tagged recombinant fraction corresponding to 137 amino acids from the internal region of human PHD finger protein 1.
Purification Method Affinity Purified
Quantity 25 μL
Regulatory Status RUO
Research Discipline Epigenetics and Nuclear Function
Primary or Secondary Primary
Test Specificity This rabbit polyclonal antibody detects Information PHD finger protein 1 (PHF1) in human cells. It targets an epitope within 137 amino acids from the internal region.
Target Species Human
Content And Storage Stable for 1 year at 2-8°C from date of receipt.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.