Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PHF10 Antibody (PCRP-PHF10-2A10), mFluor Violet 500 SE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP314153MFV500
Description
PHF10 Monoclonal antibody specifically detects PHF10 in Human samples. It is validated for Flow CytometrySpecifications
PHF10 | |
Monoclonal | |
mFluor Violet 500 SE | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified | |
RUO | |
55274 | |
Store at 4C in the dark. | |
IgG2a |
Flow Cytometry | |
PCRP-PHF10-2A10 | |
Flow Cytometry | |
BAF45a, BAF45APHD zinc finger protein XAP135, BRG1-associated factor 45a, FLJ10975, MGC111009, PHD finger protein 10, XAP135BRG1-associated factor, 45-KD, A | |
Recombinant full-length humanPHF10protein (Uniprot: Q8WUB8) | |
0.1 mL | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction