Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PHF10 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA563154
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA563154 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA563154 Supplier Invitrogen™ Supplier No. PA563154
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: GRGDEKRKNK GTSDSSSGNV SEGESPPDSQ EDSFQGRQKS KDKAATPRKD GPKRSVLSKS VPGYKPKVI Highest antigen sequence identity to the following orthologs: Mouse - 90%, Rat - 93%.

This gene contains a predicted ORF that encodes a protein with two zinc finger domains. The function of the encoded protein is not known. Sequence analysis suggests that multiple alternatively spliced transcript variants are derived from this gene but the full-length nature of only two of them is known. These two splice variants encode different isoforms. A pseudogene for this gene is located on Xq28.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PHF10
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene PHF10
Gene Accession No. Q8WUB8
Gene Alias 1810055P05Rik; AV024533; BAF45A; BRG1-associated factor 45a; BRG1-associated factor, 45-KD, A; PHD finger protein 10; PHD finger protein 10, isoform a; PHD zinc finger protein XAP135; Phf10; XAP135
Gene Symbols PHF10
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human PHF10. Recombinant protein control fragment (Product #RP-94817).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55274
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.