Learn More
Invitrogen™ PHF10 Polyclonal Antibody

Description
Immunogen sequence: PELPALDSDG DSDDGEDGRG DEKRKNKGTS DSSSGNVSEG ESPPDSQEDS For short term use, store at 2-8°C up to 1 week. For long term storage, store at -20°C in small aliquots to prevent freeze-thaw cycles. Predicted homology: Cow: 92%; Dog: 93%; Guinea Pig: 79%; Horse: 93%; Human: 100%; Mouse: 93%; Rabbit: 79%; Rat: 93%.
Specifications
Specifications
| Antigen | PHF10 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 2% sucrose and 0.09% sodium azide |
| Gene | PHF10 |
| Gene Accession No. | Q8WUB8 |
| Gene Alias | 1810055P05Rik; AV024533; BAF45A; BRG1-associated factor 45a; BRG1-associated factor, 45-KD, A; PHD finger protein 10; PHD finger protein 10, isoform a; PHD zinc finger protein XAP135; Phf10; XAP135 |
| Gene Symbols | PHF10 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.