Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PHF11 Antibody, DyLight 488, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP297698G
Description
PHF11 Polyclonal specifically detects PHF11 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
PHF11 | |
Polyclonal | |
Immunohistochemistry, Immunohistochemistry-Paraffin | |
APY, BCAPIGHER, BRCA1 C-terminus-associated protein, IgE responsiveness (atopic), IGEL, IGER, NYREN34, NY-REN-34, NY-REN-34 antigen, PHD finger protein 11, Renal carcinoma antigen NY-REN-34 | |
Produced in rabbits immunized with E. coli-derived Human PHF11 fragment. | |
0.1 mL | |
DNA replication Transcription Translation and Splicing | |
51131 | |
Store at 4°C in the dark. | |
IgG |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
DyLight 488 | |
50mM Sodium Borate | |
Rabbit | |
Antigen Affinity-purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction